Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prss3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011645.2 | NP_035775.1 | 3 | 315 | ||
GenBank mRNA | BC098498.1 | - | 3 | 298 | ||
BC141304.1 | - | 3 | 380 | 9055916 |
Gene Symbol: Gm10334 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001103153.1 | NP_001096623.1 | 3 | 331 |