Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tulp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021478.1 | NP_067453.1 | 8 | 801 | ||
GenBank mRNA | AF085681.1 | - | 8 | 849 | ||
AF105711.1 | - | 8 | 849 | |||
AK053880.1 | - | 3 | 218 | |||
AK135383.1 | - | 2 | 455 | |||
BC014820.1 | - | 8 | 807 | |||
BC029222.1 | - | 8 | 840 | 4504473 |