Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnfsf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009452.2 | NP_033478.1 | 2 | 324 | ||
GenBank mRNA | FJ176943.1 | - | 2 | 180 | ||
U12763.1 | - | 2 | 327 |