Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tyr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011661.4 | NP_035791.1 | 1 | 560 | ||
GenBank mRNA | AK014619.1 | - | 1 | 600 | ||
AK148172.1 | - | 1 | 583 | |||
AK148341.1 | - | 1 | 510 | |||
AK148357.1 | - | 1 | 583 | |||
AK148432.1 | - | 1 | 539 | |||
AY526904.1 | - | 1 | 601 | |||
BC079678.1 | - | 1 | 510 | 30539846 | ||
D00131.1 | - | 1 | 563 | |||
D00440.1 | - | 1 | 560 | |||
M20234.1 | - | 1 | 661 | |||
M24560.1 | - | 1 | 560 | |||
M26729.1 | - | 1 | 499 | |||
X12782.1 | - | 1 | 601 |