Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tyrobp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011662.2 | NP_035792.1 | 2, 3 | 148 | ||
GenBank mRNA | AF024637.1 | - | 2, 3 | 97 | ||
AF077829.1 | - | 2, 3 | 97 | |||
AK154480.1 | - | 2, 3 | 147 | |||
BC056450.1 | - | 2, 3 | 148 | 6814478 |