Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tyrobp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011662.2 | NP_035792.1 | 5 | 355 | ||
GenBank mRNA | AF024637.1 | - | 5 | 304 | ||
AF077829.1 | - | 5 | 304 | |||
BC056450.1 | - | 5 | 355 | 6814478 |