Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zrsr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011663.3 | NP_035793.1 | 1 | 2308 | ||
GenBank mRNA | AK030552.1 | - | 1 | 1098 | ||
BC132555.1 | - | 2 | 1366 | 40130832 | ||
BC138594.1 | - | 1 | 1376 | 8861614 | ||
D17407.1 | - | 2 | 2253 | |||
S69507.1 | - | 1 | 2242 |