Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ube2i | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177609.1 | NP_001171080.1 | 7, 8 | 1257 | ||
NM_001177610.1 | NP_001171081.1 | 6, 7 | 546 | |||
NM_011665.4 | NP_035795.1 | 6, 7 | 584 | |||
GenBank mRNA | AK003141.1 | - | 6, 7 | 491 | ||
AK005058.1 | - | 6, 7 | 566 | |||
AK011239.1 | - | 6, 7 | 492 | |||
AK012282.1 | - | 6, 7 | 510 | |||
AK088508.1 | - | 6, 7 | 569 | |||
AK150575.1 | - | 6, 7 | 527 | |||
AK160014.1 | - | 6, 7 | 513 | |||
AK160988.1 | - | 6, 7 | 510 | |||
AK165606.1 | - | 6, 7 | 496 | |||
AK165615.1 | - | 6, 7 | 514 | |||
AK166016.1 | - | 6, 7 | 494 | |||
AK166657.1 | - | 7, 8 | 1258 | |||
AK166930.1 | - | 6, 7 | 490 | |||
AK167162.1 | - | 7, 8 | 727 | |||
AK168212.1 | - | 6, 7 | 513 | |||
AK168706.1 | - | 6, 7 | 473 | |||
AK168766.1 | - | 6, 7 | 507 | |||
U31934.1 | - | 5, 6 | 424 | |||
U76416.1 | - | 6, 7 | 520 | |||
U82627.1 | - | 6, 7 | 529 | |||
U94402.1 | - | 6, 7 | 580 | |||
X97575.1 | - | 6, 7 | 461 | |||
X99739.1 | - | 6, 7 | 508 |