Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc35a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083937.1 | NP_001077406.1 | 4 | 962 | ||
NM_078484.2 | NP_511039.2 | 4 | 962 | |||
GenBank mRNA | AB027147.1 | - | 4 | 881 | ||
AF229634.1 | - | 4 | 962 | |||
AK146830.1 | - | 4 | 878 | |||
AK155755.1 | - | 4 | 874 | |||
BC037701.1 | - | 4 | 899 | 4187325 |