Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nr1h3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177730.1 | NP_001171201.1 | 4 | 644 | ||
NM_013839.4 | NP_038867.2 | 4 | 797 | |||
GenBank mRNA | AF085745.1 | - | 4 | 539 | ||
AJ132601.1 | - | 4 | 575 | |||
AK143904.1 | - | 4 | 578 | |||
AK159250.1 | - | 4 | 798 | |||
AY195870.1 | - | 4 | 644 | |||
AY195871.1 | - | 4 | 644 | |||
BC012646.1 | - | 4 | 585 | 4163109 |