Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kdm6a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009483.1 | NP_033509.1 | 16 | 2188 | ||
GenBank mRNA | AJ002730.1 | - | 15 | 1647 | ||
AK080370.1 | - | 16 | 2207 | |||
AK144654.1 | - | 14 | 2024 | |||
BC027130.1 | - | 4 | 635 | |||
BC053433.1 | - | 16 | 2188 | 6516368 | ||
BC075703.1 | - | 7 | 1098 |