Gene Symbol: ITGB5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002213.4 | NP_002204.2 | 13 | 2382 | ||
XM_005247436.2 | XP_005247493.1 | 12 | 2525 | |||
XM_006713630.2 | XP_006713693.1 | 12 | 2480 | |||
XM_017006351.1 | XP_016861840.1 | 13 | 2153 | |||
XM_017006352.1 | XP_016861841.1 | 13 | 2092 | |||
XM_017006353.1 | XP_016861842.1 | 13 | 2124 | |||
XM_017006354.1 | XP_016861843.1 | 10 | 2267 | |||
XM_017006355.1 | XP_016861844.1 | 12 | 2002 | |||
GenBank mRNA | AK054968.1 | - | 2 | 1230 | ||
AK309551.1 | - | 5 | 868 | |||
AK315429.1 | - | 13 | 2118 | |||
BC006541.2 | - | 13 | 2374 | 2958666 | ||
J05633.1 | - | 13 | 2123 | |||
M35011.1 | - | 13 | 2106 | |||
X53002.1 | - | 13 | 2413 |