Gene Symbol: ITGB5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002213.4 | NP_002204.2 | 7 | 1334 | ||
XM_005247436.2 | XP_005247493.1 | 7 | 1567 | |||
XM_006713630.2 | XP_006713693.1 | 7 | 1567 | |||
XM_017006351.1 | XP_016861840.1 | 7 | 1105 | |||
XM_017006352.1 | XP_016861841.1 | 7 | 1044 | |||
XM_017006353.1 | XP_016861842.1 | 7 | 1076 | |||
XM_017006355.1 | XP_016861844.1 | 7 | 1044 | |||
GenBank mRNA | AK315429.1 | - | 7 | 1070 | ||
BC006541.2 | - | 7 | 1326 | 2958666 | ||
J05633.1 | - | 7 | 1075 | |||
M35011.1 | - | 7 | 1058 | |||
X53002.1 | - | 7 | 1365 |