Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Vegfb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001185164.1 | NP_001172093.1 | 7 | 1114 | ||
NM_011697.3 | NP_035827.1 | 7 | 1215 | |||
GenBank mRNA | AK148188.1 | - | 7 | 1214 | ||
AK210361.1 | - | 1 | 280 | |||
BC046303.1 | - | 7 | 1092 | 6492277 | ||
CF804631.1 | - | 1 | 129 | |||
U43836.1 | - | 7 | 1167 |