Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vipr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011703.4 | NP_035833.2 | 8 | 974 | ||
GenBank mRNA | AF266282.1 | - | 8 | 856 | ||
AK052465.1 | - | 8 | 975 |
Set Membership: |
![]() |