Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vwf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011708.4 | NP_035838.3 | 15 | 2078 | ||
GenBank mRNA | AF539800.2 | - | 14 | 1826 | ||
AK044921.1 | - | 15 | 2066 | |||
AK157296.1 | - | 15 | 2080 | |||
AK170201.1 | - | 2 | 438 | |||
AY208897.1 | - | 15 | 1886 | |||
DQ355287.1 | - | 15 | 2063 | |||
DQ355288.1 | - | 15 | 2063 |