Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wnt10b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011718.2 | NP_035848.1 | 5 | 1265 | ||
GenBank mRNA | AK029284.1 | - | 4 | 708 | ||
BC114969.1 | - | 6 | 1045 | |||
L33719.1 | - | 1 | 200 | |||
U20658.1 | - | 5 | 1080 | |||
U30464.1 | - | 5 | 1263 | |||
U61970.1 | - | 4 | 998 | |||
U61971.1 | - | 4 | 710 |