Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wnt8b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011720.3 | NP_035850.2 | 3 | 278 | ||
GenBank mRNA | AF130349.1 | - | 3 | 280 | ||
AK050992.1 | - | 3 | 279 | |||
BC119544.2 | - | 3 | 210 | 40056929 |