Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zan | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011741.2 | NP_035871.2 | 48 | 9538 | ||
GenBank mRNA | U97068.1 | - | 50 | 9406 |