Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Zap70 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289612.1 | NP_001276541.1 | 5 | 951 | ||
NM_001289765.1 | NP_001276694.1 | 12 | 1898 | |||
NM_001289766.1 | NP_001276695.1 | 12 | 1863 | |||
NM_009539.3 | NP_033565.2 | 12 | 1855 | |||
GenBank mRNA | AB083210.1 | - | 5 | 954 | ||
AB084383.1 | - | 5 | 793 | |||
AK039883.1 | - | 12 | 1792 | |||
AK089056.1 | - | 12 | 1827 | |||
BC029727.1 | - | 12 | 1813 | 4925739 | ||
U04379.1 | - | 12 | 1809 | |||
U77667.1 | - | 11 | 1701 |