Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037707.1 | NP_001032796.1 | 5 | 1573 | ||
NM_001285797.1 | NP_001272726.1 | 5 | 1845 | |||
NM_001285798.1 | NP_001272727.1 | 5 | 1799 | |||
NM_011754.2 | NP_035884.2 | 5 | 1568 | |||
GenBank mRNA | AK028500.1 | - | 5 | 1800 | ||
AK135293.1 | - | 5 | 1849 | |||
AK147732.1 | - | 5 | 1563 | |||
BC059037.1 | - | 3 | 1512 | 6812360 | ||
M36515.1 | - | 2 | 445 | |||
X12593.1 | - | 2 | 445 |