Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Zfp41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001044718.2 | NP_001038183.1 | 2 | 642 | ||
NM_011759.3 | NP_035889.1 | 2 | 850 | |||
GenBank mRNA | AK030420.1 | - | 2 | 812 | ||
BC053927.1 | - | 2 | 567 | 6314215 | ||
D10631.1 | - | 1 | 84 |