Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009557.3 | NP_033583.2 | 2 | 609 | ||
GenBank mRNA | AK051031.1 | - | 2 | 582 | ||
AK053652.1 | - | 2 | 612 | |||
AK136405.1 | - | 2 | 518 | |||
AK148255.1 | - | 2 | 558 | |||
AK173240.1 | - | 2 | 596 | |||
BC006587.1 | - | 2 | 276 | 3587306 |