Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009557.3 | NP_033583.2 | 3 | 766 | ||
GenBank mRNA | AK051031.1 | - | 3 | 739 | ||
AK053652.1 | - | 3 | 769 | |||
AK136405.1 | - | 3 | 675 | |||
AK148255.1 | - | 3 | 715 | |||
AK173240.1 | - | 3 | 753 | |||
BC006587.1 | - | 3 | 433 | 3587306 | ||
BC092398.1 | - | 1 | 76 | |||
M98502.1 | - | 1 | 289 |