Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp62 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024846.1 | NP_001020017.1 | 5 | 1647 | ||
NM_009562.2 | NP_033588.2 | 4 | 1547 | |||
GenBank mRNA | AK028351.1 | - | 5 | 1650 | ||
AK048619.1 | - | 4 | 1549 | |||
AK088060.1 | - | 4 | 1725 | |||
AK154767.1 | - | 4 | 1538 | |||
AK157730.1 | - | 4 | 1631 | |||
AK165654.1 | - | 4 | 1529 | |||
AK167091.1 | - | 4 | 1565 | |||
BC022935.1 | - | 4 | 1665 | 3497847 | ||
BC065692.1 | - | 2 | 1751 | 6826605 | ||
L36315.1 | - | 501 | ||||
Z67747.1 | - | 1 | 200 |