Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Clec5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038604.1 | NP_001033693.1 | 5 | 458 | ||
NM_021364.2 | NP_067339.1 | 4 | 383 | |||
GenBank mRNA | AF139769.1 | - | 4 | 353 | ||
AK036697.1 | - | 4 | 349 | |||
AK046600.1 | - | 4 | 370 | |||
AK137482.1 | - | 4 | 384 | |||
BC104364.1 | - | 5 | 417 | 40049817 | ||
BC104365.1 | - | 5 | 417 | 40049818 | ||
CN670295.1 | - | 5 | 420 |