Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dlg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011807.3 | NP_035937.2 | 8, 9 | 1102 | ||
GenBank mRNA | AF388675.1 | - | 8, 9 | 881 | ||
AK046525.1 | - | 7, 8 | 1142 |