Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ets2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011809.3 | NP_035939.3 | 7 | 981 | ||
GenBank mRNA | AK014602.2 | - | 7 | 903 | ||
AK030537.1 | - | 3 | 283 | |||
AK038348.1 | - | 7 | 968 | |||
AK128933.1 | - | 8 | 855 | |||
AK143687.1 | - | 7 | 933 | |||
AK144974.1 | - | 7 | 932 | |||
AK145852.1 | - | 7 | 817 | |||
AK150175.1 | - | 7 | 910 | |||
AK151315.1 | - | 7 | 932 | |||
AK152750.1 | - | 7 | 935 | |||
AK153339.1 | - | 7 | 933 | |||
AK153478.1 | - | 7 | 933 | |||
BC005486.1 | - | 7 | 902 | 3486021 | ||
BC005504.1 | - | 7 | 842 | 3490813 | ||
CT010235.1 | - | 6 | 743 | |||
J04103.1 | - | 7 | 880 |