Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krtap14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013707.2 | NP_038735.2 | 1 | 424 | ||
GenBank mRNA | AF003691.1 | - | 1 | 413 | ||
BC104262.1 | - | 1 | 398 | 40040069 | ||
BC104263.1 | - | 1 | 397 | 40040071 | ||
D85925.1 | - | 1 | 413 |