Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oasl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011854.2 | NP_035984.2 | 2 | 1171 | ||
GenBank mRNA | AF068835.1 | - | 2 | 1157 | ||
AK010034.1 | - | 2 | 513 | |||
AK150492.1 | - | 2 | 502 | |||
AK157649.1 | - | 2 | 537 | |||
BC034361.1 | - | 2 | 598 | 2646375 |