Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pebp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018858.2 | NP_061346.2 | 2 | 251 | ||
GenBank mRNA | AB046417.1 | - | 2 | 253 | ||
AF300422.1 | - | 2 | 230 | |||
AK008037.1 | - | 267 | ||||
AK088212.1 | - | 2 | 256 | |||
AK143897.1 | - | 2 | 283 | |||
AK149815.1 | - | 2 | 267 | |||
AK159842.1 | - | 2 | 256 | |||
AK182328.1 | - | 2 | 141 | |||
BC008169.1 | - | 2 | 285 | 3257403 | ||
BC083063.1 | - | 2 | 251 | 5362125 | ||
BC089332.1 | - | 2 | 234 | 30359654 | ||
U43206.1 | - | 2 | 249 |