Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptpn21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146199.1 | NP_001139671.1 | 16 | 3120 | ||
NM_011877.2 | NP_036007.2 | 16 | 3255 | |||
GenBank mRNA | BC049893.1 | - | 14 | 3002 | ||
BC055942.1 | - | 4 | 1644 | |||
BC066856.1 | - | 14 | 1514 | |||
BC098500.1 | - | 16 | 3120 | 30081068 | ||
D37801.1 | - | 16 | 3204 | |||
D83072.1 | - | 3 | 516 |