Search Thermo Fisher Scientific
Gene Symbol: Spry4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011898.2 | NP_036028.2 | 2 | 258 | ||
GenBank mRNA | AB019280.1 | - | 2 | 327 | ||
AF176906.1 | - | 2 | 411 | |||
AK037180.1 | - | 2 | 333 | |||
AK048122.1 | - | 2 | 336 | |||
AK139816.1 | - | 2 | 317 | |||
BC050944.1 | - | 2 | 258 | 6821944 | ||
BC057005.1 | - | 2 | 258 | 6821944 |