Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clcn6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011929.2 | NP_036059.1 | 16 | 1678 | ||
GenBank mRNA | AK145053.1 | - | 16 | 1723 | ||
AK165362.1 | - | 16 | 1728 | |||
AK220319.1 | - | 10 | 1168 |