Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clcn7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011930.3 | NP_036060.1 | 15 | 1362 | ||
GenBank mRNA | AK030444.1 | - | 15 | 1363 | ||
AK032674.1 | - | 2 | 92 | |||
AK128981.1 | - | 15 | 1330 | |||
AK150395.1 | - | 15 | 1273 | |||
AK197679.1 | - | 1 | 61 | |||
AK203333.1 | - | 1 | 63 | |||
BC050907.1 | - | 15 | 1334 | 5698121 | ||
BC053049.1 | - | 15 | 1334 | 5698121 | ||
BC054799.1 | - | 15 | 1342 | 6849205 |