Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grk6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038018.4 | NP_001033107.1 | 6 | 677 | ||
NM_001112711.2 | NP_001106182.1 | 6 | 677 | |||
NM_001286063.1 | NP_001272992.1 | 6 | 657 | |||
NM_001286064.1 | NP_001272993.1 | 6 | 677 | |||
NM_001286065.1 | NP_001272994.1 | 6 | 677 | |||
NM_001286066.1 | NP_001272995.1 | 6 | 677 | |||
NM_011938.4 | NP_036068.2 | 6 | 677 | |||
GenBank mRNA | AF040747.1 | - | 6 | 547 | ||
AF040748.1 | - | 6 | 547 | |||
AF040749.1 | - | 6 | 547 | |||
AK051405.1 | - | 6 | 659 | |||
AK146119.1 | - | 6 | 552 | |||
AK161142.1 | - | 4 | 309 | |||
BC057206.1 | - | 6 | 666 | |||
BC075719.1 | - | 6 | 558 | 5031455 | ||
Y15797.1 | - | 6 | 499 | |||
Y15798.1 | - | 6 | 521 | |||
Y15799.1 | - | 6 | 504 | |||
Y15800.1 | - | 6 | 499 |