Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mapk10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081567.1 | NP_001075036.1 | 8 | 1224 | ||
NM_009158.2 | NP_033184.2 | 8 | 1224 | |||
GenBank mRNA | AB005665.1 | - | 5 | 607 | ||
AB096076.1 | - | 6 | 658 | |||
AB096077.1 | - | 6 | 658 | |||
AB096078.1 | - | 6 | 658 | |||
AB096079.1 | - | 6 | 658 | |||
AK036275.1 | - | 8 | 1176 | |||
AK042368.1 | - | 7 | 789 | |||
AK082242.1 | - | 8 | 1228 | |||
AK163206.1 | - | 8 | 1175 | |||
AK163385.1 | - | 5 | 701 | |||
BC046625.1 | - | 8 | 1163 | 6494163 | ||
L35236.1 | - | 8 | 967 |