Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Irf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252382.1 | NP_001239311.1 | 3 | 492 | ||
NM_012057.4 | NP_036187.1 | 3 | 440 | |||
GenBank mRNA | AF028725.1 | - | 3 | 332 | ||
AK150598.1 | - | 3 | 361 | |||
AK151019.1 | - | 3 | 357 | |||
AK152042.1 | - | 3 | 361 | |||
AK156220.1 | - | 3 | 492 | |||
AK171356.1 | - | 3 | 360 | |||
BY749561.1 | - | 3 | 440 | |||
BY749804.1 | - | 3 | 356 | |||
EU401974.1 | - | 2 | 279 |