Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Azi2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001048146.2 | NP_001041611.1 | 7 | 1184 | ||
NM_001286507.1 | NP_001273436.1 | 6 | 1084 | |||
NM_001286508.1 | NP_001273437.1 | 7 | 1102 | |||
NM_013727.4 | NP_038755.1 | 7 | 1184 | |||
GenBank mRNA | AB007141.1 | - | 7 | 1020 | ||
AK004992.1 | - | 7 | 1163 | |||
AK015942.1 | - | 7 | 1005 | |||
AK030076.1 | - | 7 | 1016 | |||
AK036102.1 | - | 6 | 991 | |||
AK134714.1 | - | 7 | 1087 | |||
AK153477.1 | - | 7 | 1148 | |||
AK165549.1 | - | 7 | 1070 | |||
AK167783.1 | - | 7 | 1128 | |||
BC006851.1 | - | 7 | 1079 | 3156857 |