Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nbn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013752.3 | NP_038780.3 | 9 | 1189 | ||
GenBank mRNA | AB016988.1 | - | 9 | 1111 | ||
AF076687.1 | - | 9 | 1183 | |||
AF092840.1 | - | 9 | 1172 | |||
AK031933.1 | - | 9 | 1175 | |||
AK132302.1 | - | 9 | 1230 | |||
AK134960.1 | - | 9 | 1217 | |||
AK159790.1 | - | 9 | 1196 | |||
AK160614.1 | - | 9 | 1156 | |||
BC003719.1 | - | 9 | 1198 | |||
BC013190.1 | - | 9 | 1189 | |||
BC044773.2 | - | 9 | 1188 | 5067851 | ||
BC055061.1 | - | 9 | 1181 | 6528890 |