Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Add3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164099.2 | NP_001157571.1 | 15 | 2199 | ||
NM_001164100.2 | NP_001157572.1 | 14 | 2165 | |||
NM_001164101.2 | NP_001157573.1 | 14 | 2103 | |||
NM_001277100.1 | NP_001264029.1 | 15 | 2261 | |||
NM_013758.4 | NP_038786.2 | 16 | 2011 | |||
GenBank mRNA | AF100424.1 | - | 13 | 1834 | ||
AF100425.1 | - | 14 | 1930 | |||
AF189772.1 | - | 5 | 614 | |||
AK019491.1 | - | 15 | 2140 | |||
AK028481.1 | - | 15 | 2104 | |||
AK035151.1 | - | 14 | 1932 | |||
AK083939.1 | - | 14 | 2006 | |||
AK144174.1 | - | 14 | 2109 | |||
BC004032.1 | - | 14 | 1882 | |||
BC004746.1 | - | 8 | 1009 | |||
BC029196.1 | - | 15 | 2064 | |||
BC037116.1 | - | 16 | 2022 | 4506748 | ||
BC060097.1 | - | 15 | 2044 |