Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh2d3c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252547.1 | NP_001239476.1 | 10 | 1824 | ||
NM_013781.3 | NP_038809.1 | 11 | 2310 | |||
GenBank mRNA | AB030442.1 | - | 10 | 1787 | ||
AB043953.1 | - | 11 | 2293 | |||
AF168364.1 | - | 11 | 2251 | |||
AK042709.1 | - | 10 | 1808 | |||
AK085885.1 | - | 1808 | ||||
AK155165.1 | - | 11 | 2246 | |||
AK172072.1 | - | 12 | 2689 | |||
BC113203.1 | - | 11 | 2265 | 40048064 |