Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Nagpa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013796.3 | NP_038824.2 | 2, 3 | 568 | ||
GenBank mRNA | AK032158.1 | - | 2, 3 | 569 | ||
AK087468.1 | - | 2, 3 | 568 | |||
AK138035.1 | - | 2, 3 | 561 | |||
AK155450.1 | - | 2, 3 | 547 | |||
AK159629.1 | - | 2, 3 | 547 | |||
AK162633.1 | - | 2, 3 | 550 | |||
AK168830.1 | - | 1, 2 | 655 | |||
AK170219.1 | - | 2, 3 | 550 | |||
AK211874.1 | - | 1, 2 | 83 | |||
BC019640.1 | - | 1, 2 | 630 | |||
BC039790.1 | - | 2, 3 | 560 | 4951805 |