Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Shroom3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077595.2 | NP_001071063.1 | 9, 10 | 5441 | ||
NM_001077596.2 | NP_001071064.1 | 9, 10 | 5489 | |||
NM_015756.2 | NP_056571.2 | 10, 11 | 5813 | |||
GenBank mRNA | AF199421.1 | - | 10, 11 | 5831 | ||
AF199422.1 | - | 9, 10 | 5399 | |||
AK049347.1 | - | 6, 7 | 3347 | |||
AK129371.1 | - | 6, 7 | 4819 | |||
AK147493.1 | - | 9, 10 | 5424 | |||
AK164918.1 | - | 9, 10 | 5400 | |||
BC003909.1 | - | 4, 5 | 1085 | |||
BC145113.1 | - | 11, 12 | 5230 | 9053661 |