Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nelfe | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001045863.1 | NP_001039328.1 | 8, 9 | 1089 | ||
NM_001045864.1 | NP_001039329.1 | 8, 9 | 1175 | |||
NM_138580.2 | NP_613046.2 | 8, 9 | 963 | |||
GenBank mRNA | AK011663.1 | - | 8, 9 | 948 | ||
AK012389.1 | - | 8, 9 | 980 | |||
AK131947.1 | - | 8, 9 | 947 | |||
AK147011.1 | - | 8, 9 | 1071 | |||
BC034867.1 | - | 8, 9 | 915 | 1495547 | ||
DV662511.1 | - | 5, 6 | 708 |