Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other KCNMA1 SIRNA Assays ›
Gene Symbol:
KCNMA1
Gene Name:
potassium calcium-activated channel subfamily M alpha 1
Gene Aliases:
BKTM, KCa1.1, MaxiK, SAKCA, SLO, SLO-ALPHA, SLO1, bA205K10.1, hSlo, mSLO1
Chromosome Location:
Chr.10: 76869601 - 77637819 on Build GRCh38
UniGene ID:
Hs.144795
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s7788
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: KCNMA1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001014797.2
NP_001014797.1
1
250
NM_001161352.1
NP_001154824.1
1
250
NM_001161353.1
NP_001154825.1
1
250
NM_001271518.1
NP_001258447.1
1
250
NM_001271519.1
NP_001258448.1
1
250
NM_001271520.1
NP_001258449.1
1
250
NM_001271521.1
NP_001258450.1
1
250
NM_001271522.1
NP_001258451.1
1
250
NM_001322829.1
NP_001309758.1
1
250
NM_001322830.1
NP_001309759.1
1
250
NM_001322832.1
NP_001309761.1
1
250
NM_001322835.1
NP_001309764.1
1
250
NM_001322836.1
NP_001309765.1
1
250
NM_001322837.1
NP_001309766.1
1
250
NM_001322839.1
NP_001309768.1
1
250
NM_002247.3
NP_002238.2
1
250
XM_005269776.3
XP_005269833.1
1
239
XM_005269778.1
XP_005269835.1
1
239
XM_005269781.2
XP_005269838.1
1
171
XM_005269787.3
XP_005269844.1
1
239
XM_005269789.2
XP_005269846.1
1
171
XM_005269792.2
XP_005269849.1
1
171
XM_005269796.2
XP_005269853.1
1
171
XM_006717826.2
XP_006717889.1
1
171
XM_011539773.1
XP_011538075.1
1
239
XM_011539774.1
XP_011538076.1
1
239
XM_011539775.1
XP_011538077.1
1
239
XM_011539776.1
XP_011538078.1
1
239
XM_011539777.1
XP_011538079.1
1
239
XM_011539778.2
XP_011538080.1
1
171
XM_011539779.1
XP_011538081.1
1
239
XM_011539780.1
XP_011538082.1
1
239
XM_011539781.2
XP_011538083.1
1
239
XM_011539782.2
XP_011538084.1
1
191
XM_011539783.1
XP_011538085.1
1
239
XM_011539784.1
XP_011538086.1
1
239
XM_011539785.1
XP_011538087.1
1
239
XM_017016207.1
XP_016871696.1
1
239
XM_017016208.1
XP_016871697.1
1
239
XM_017016209.1
XP_016871698.1
1
239
XM_017016210.1
XP_016871699.1
1
239
XM_017016211.1
XP_016871700.1
1
239
XM_017016212.1
XP_016871701.1
1
239
XM_017016213.1
XP_016871702.1
1
239
XM_017016214.1
XP_016871703.1
1
239
XM_017016215.1
XP_016871704.1
1
239
XM_017016216.1
XP_016871705.1
1
239
XM_017016217.1
XP_016871706.1
1
191
XM_017016219.1
XP_016871708.1
1
239
XM_017016220.1
XP_016871709.1
1
239
XM_017016221.1
XP_016871710.1
1
239
XM_017016222.1
XP_016871711.1
1
239
GenBank mRNA
AB113382.1
-
1
15
AB524033.1
-
1
73
AY040849.1
-
1
15
BC062659.1
-
1
245
BC137115.1
-
1
153
9021112
BC137137.1
-
1
153
9021134
BC144496.1
-
1
153
9053023
CR627384.1
-
1
156
U11717.1
-
1
25
U13913.1
-
1
151
U23767.1
-
1
73
Back To Top
More Information
Panther Classification:
Molecular Function -
ion channel
Gene Ontology Categories:
Function(s)
Process(es)
response to hypoxia
potassium ion transport
response to osmotic stress
cellular potassium ion homeostasis
response to carbon monoxide
regulation of ion transmembrane transport
regulation of membrane potential
positive regulation of apoptotic process
negative regulation of cell volume
response to calcium ion
micturition
smooth muscle contraction involved in micturition
potassium ion transmembrane transport
actin binding
voltage-gated potassium channel activity
protein binding
calcium-activated potassium channel activity
metal ion binding
large conductance calcium-activated potassium channel activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-8gqds:80/10.239.164.183:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0