Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: D10Wsu102e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026579.3 | NP_080855.1 | 4 | 509 | ||
GenBank mRNA | AK020067.1 | - | 4 | 487 | ||
AK049544.1 | - | 4 | 492 | |||
AK128995.1 | - | 4 | 502 | |||
AK147063.1 | - | 4 | 492 | |||
AK160350.1 | - | 4 | 492 | |||
AK162255.1 | - | 4 | 485 | |||
AK199731.1 | - | 2 | 155 | |||
BC035946.1 | - | 4 | 466 | |||
BC050904.1 | - | 4 | 477 | 6416625 | ||
BC094241.1 | - | 4 | 471 | 30610591 |