Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030685.3 | NP_109610.1 | 3 | 1314 | ||
GenBank mRNA | AB041655.1 | - | 3 | 1183 | ||
AK002653.1 | - | 3 | 1186 | |||
AK032680.1 | - | 3 | 1184 | |||
AK050508.1 | - | 3 | 1154 | |||
AK075776.1 | - | 3 | 1180 | |||
AK144732.1 | - | 3 | 1179 | |||
AK148485.1 | - | 3 | 1185 | |||
AK149786.1 | - | 3 | 1184 | |||
AK150005.1 | - | 3 | 1184 | |||
AK150025.1 | - | 3 | 1184 | |||
AK150368.1 | - | 3 | 1183 | |||
AK150428.1 | - | 3 | 1184 | |||
AK150659.1 | - | 3 | 1179 | |||
AK150714.1 | - | 3 | 1183 | |||
AK150809.1 | - | 3 | 1153 | |||
AK151323.1 | - | 3 | 1153 | |||
AK151384.1 | - | 3 | 1166 | |||
AK151669.1 | - | 3 | 1179 | |||
AK151853.1 | - | 3 | 1182 | |||
AK152373.1 | - | 3 | 1179 | |||
AK152383.1 | - | 3 | 1183 | |||
AK152583.1 | - | 3 | 1179 | |||
AK152636.1 | - | 3 | 1184 | |||
AK152806.1 | - | 3 | 1184 | |||
AK152882.1 | - | 3 | 1179 | |||
AK152996.1 | - | 3 | 1185 | |||
AK153056.1 | - | 3 | 1153 | |||
AK153118.1 | - | 3 | 1153 | |||
AK153138.1 | - | 3 | 1179 | |||
AK153286.1 | - | 3 | 1153 | |||
AK153415.1 | - | 3 | 1183 | |||
AK153456.1 | - | 2 | 836 | |||
AK159271.1 | - | 3 | 1184 | |||
AK161595.1 | - | 3 | 1184 | |||
AK166004.1 | - | 3 | 1179 | |||
AK166052.1 | - | 3 | 1159 | |||
AK167482.1 | - | 3 | 1155 | |||
AK167952.1 | - | 3 | 1184 | |||
AK168046.1 | - | 3 | 1184 | |||
AK168569.1 | - | 3 | 1183 | |||
AK168734.1 | - | 3 | 1183 | |||
AK168754.1 | - | 3 | 1183 | |||
AK168947.1 | - | 3 | 1183 | |||
AK169049.1 | - | 3 | 1179 | |||
AK169175.1 | - | 3 | 1184 | |||
AK170734.1 | - | 3 | 1184 | |||
BC002114.1 | - | 3 | 1160 | 3489738 |