Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spdef | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013891.4 | NP_038919.1 | 2 | 820 | ||
GenBank mRNA | AB019436.1 | - | 2 | 756 | ||
AK078863.1 | - | 2 | 821 | |||
BC012648.1 | - | 2 | 821 | 4218741 |