Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kdm2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013910.2 | NP_038938.1 | 12 | 2430 | ||
GenBank mRNA | AF176524.2 | - | 5 | 851 | ||
AK012952.1 | - | 2 | 297 | |||
AK031626.1 | - | 12 | 2396 | |||
AK129479.1 | - | 23 | 3867 | |||
BC057622.1 | - | 12 | 2425 | 6402440 | ||
BC065090.1 | - | 23 | 4066 | 6851421 |